missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | MYBPC2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628132
|
Novus Biologicals
NBP2-93972-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655192
|
Novus Biologicals
NBP2-93972-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYBPC2 Polyclonal antibody specifically detects MYBPC2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MYBPC2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4606 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C-protein, skeletal muscle fast isoform, fast MyBP-C, fast-type muscle myosin-binding-protein C, MGC163408, MYBPC, MYBPCF, myosin binding protein C, fast type, myosin-binding protein C, fast-type | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 912-1141 of human MYBPC2 (NP_004524.3). LSVQIENMKDTATIRIRVVEKAGPPINVMVKEVWGTNALVEWQAPKDDGNSEIMGYFVQKADKKTMEWFNVYERNRHTSCTVSDLIVGNEYYFRVYTENICGLSDSPGVSKNTARILKTGITFKPFEYKEHDFRMAPKFLTPLIDRVVVAGYSAALNCAVRGHPKPKVVWMKNKMEIREDPKFLITNYQGVLTLNIRRPSPFDAGTYTCRAVNELGEALAECKLEVRVPQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title