missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC2 Monoclonal Antibody (OTI2E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811203
This item is not returnable.
View return policy
Description
Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the fast-type isoform designated MYBPC2. Both the fast-type and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. MYBPC2 is typically required for strong contractions and functions under anaerobic conditions. It is more similar to the cardiac isoform (MYBPC3) than to the slow-type isoform (MYBPC1) in terms of pCa50-indexed force development, length-independent cooperativity and length dependent activation. It has been suggested that in cardiac and fast muscle MYBPC2 contributes to an internal load, possibly by binding to actin via its N-terminal region.Specifications
| MYBPC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14324 | |
| MYBPC2 | |
| Full length human recombit protein of human MYBPC2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2E1 | |
| Unconjugated | |
| MYBPC2 | |
| MYBPC, MYBPCF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering