missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC2 Monoclonal Antibody (OTI2E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811203S
This item is not returnable.
View return policy
Description
MYBPC2 Monoclonal antibody specifically detects MYBPC2 in Human samples. It is validated for Western BlotSpecifications
| MYBPC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15119 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6926 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2E1 | |
| Unconjugated | |
| Tbx3 | |
| C-protein, skeletal muscle fast isoform; Fast MyBP-C; fast-type C-protein; fast-type muscle myosin-binding-protein C; MYBPC; Mybpc2; MYBPCF; myosin binding protein C, fast type; myosin binding protein C, fast-type; myosin-binding protein C, fast-type; mypc2; testicular tissue protein Li 126 | |
| Human recombinant protein fragment corresponding to amino acids 244-481 of human TBX3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction