missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC3 Mouse anti-Human, Clone: OTI6F6, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813814S
This item is not returnable.
View return policy
Description
MYBPC3 Monoclonal antibody specifically detects MYBPC3 in Human samples. It is validated for ELISASpecifications
| MYBPC3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14896 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4607 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI6F6 | |
| Unconjugated | |
| MYBPC3 | |
| cardiac C-protein; cardiac MyBP-C; cardiac myosin binding protein C; CMD1MM; CMH4; C-protein, cardiac muscle isoform; DKFZp779E1762; FHC; LVNC10; MYBP-C; Mybpc3; myosin binding protein C, cardiac; myosin binding protein C3; myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C | |
| Full length human recombinant protein of human MYBPC3 (NP_000247) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction