Learn More
MYBPC3 Mouse anti-Human, Unconjugated, Clone: 3C8F8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67608-1-IG-150UL
Description
The MYBPC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYBPC3. This antibody recognizes human, mouse, rat, pig antigen. The MYBPC3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MYBPC3 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry | |
| 3C8F8 | |
| Unconjugated | |
| 150 μL | |
| 4607 | |
| IgG1 |