missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYBPC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19977-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody detects the full length form and the degradation bands around 28-54 kDa in SDS-PAGE.
MYBPC3 encodes the cardiac isoform of myosin-binding protein C. Myosin-binding protein C is a myosin-associated protein found in the cross-bridge-bearing zone (C region) of A bands in striated muscle. MYBPC3, the cardiac isoform, is expressed exclussively in heart muscle. Regulatory phosphorylation of the cardiac isoform in vivo by cAMP-dependent protein kinase (PKA) upon adrenergic stimulation may be linked to modulation of cardiac contraction. Mutations in MYBPC3 are one cause of familial hypertrophic cardiomyopathy.Specifications
| MYBPC3 | |
| Polyclonal | |
| Unconjugated | |
| MYBPC3 | |
| Cardiac MyBP C, CMH4, FHC, MYBP C, MYBPC3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17868, 295929, 4607 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70468, P56741, Q14896 | |
| Mybpc3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction