Learn More
MYCN Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809361S
Encompass_Preferred
 View more versions of this product
Product Code. 17561514
  • € 242.00 / 30µL
Description
MYCN Monoclonal antibody specifically detects MYCN in Human samples. It is validated for Western Blot
Specifications
Specifications
MYCN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14520
Mouse
Affinity chromatography
RUO
3026
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1F1
Unconjugated
HABP2
Avian myelocytomatosis viral (v-myc) related oncogene neuroblastoma derived (Nmyc); Avian myelocytomatosis viral (v-myc) related oncogene, neuroblastoma derived (Nmyc); BHLHE37; Class E basic helix-loop-helix protein 37; c-nmyc; fb57a02; MODED; Mycn; mycn protein; MYCN proto-oncogene, bHLH transcription factor; neuroblastoma MYC oncogene; neuroblastoma myc-related oncogene 1; neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene; Nmuc1; NMYC; N-myc; N-myc protein; N-myc proto-oncogene protein; Nmyc1; Nmyc-1; N-myc-like protein; ODED; oncogene NMYC; oncogene pp65/67; pp65/67; v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian); wu:fb57a02; zgc:85706
Human recombinant protein fragment corresponding to amino acids 33-330 of human HABP2 produced in E.coli.