missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium avium Antibody (2A4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64559AF350
This item is not returnable.
View return policy
Description
Mycobacterium avium Monoclonal specifically detects Mycobacterium avium in Bacteria samples. It is validated for ELISA.
Specifications
| Mycobacterium avium | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Bacteria | |
| IgG1 |
| ELISA | |
| 2A4 | |
| ELISA | |
| m avium, M. avium | |
| Mycobacterium Avium cell extract. | |
| 0.1 mL | |
| Specific for Mycobacterium avium. It does not react with Mycobacterium tuberculosis, Mycobacterium phlei, Corynebacterium bovis or Mycoplasma bovis. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction