missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium bovis Antibody (11G3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 521.00
Specifications
| Antigen | Mycobacterium bovis |
|---|---|
| Clone | 11G3 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 |
| Applications | ELISA, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612768
|
Novus Biologicals
NBP2-54631-0.025mg |
0.025 mg |
€ 214.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668866
|
Novus Biologicals
NBP2-54631 |
0.1 mg |
€ 521.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mycobacterium bovis Monoclonal antibody specifically detects Mycobacterium bovis in Bacteria samples. It is validated for ELISA, ImmunoprecipitationSpecifications
| Mycobacterium bovis | |
| 1 mg/mL | |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Bacteria | |
| Bacterium tuberculosis, Mycobacterium africanum, Mycobacterium bovis, Mycobacterium bovis subsp. caprae, Mycobacterium caprae, Mycobacterium microti, Mycobacterium pinnipedii, Mycobacterium tuberculosis subsp. caprae | |
| IgM | |
| Thiophilic Chromatography |
| 11G3 | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS | |
| Gamma-irradiated whole M. bovis AF2122/97 cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title