missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium tuberculosis Antibody (BGN/1209/3875), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62769H
This item is not returnable.
View return policy
Description
Mycobacterium tuberculosis Monoclonal antibody specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
| Mycobacterium tuberculosis | |
| Monoclonal | |
| HRP | |
| M. tuberculosis | |
| Recombinant 38 kDa antigen from Mycobacterium tuberculosis. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, ELISA | |
| BGN/1209/3875 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction