Learn More
Mycobacterium Tuberculosis CFP10 Antibody (KFB42), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-05467UV
Product Code. 18462095
  • ONLINE EXCLUSIVE PRICE
    € 452.00
    € 427.35 / 0.20mL
    Save € 24.65
    5% Off
Description
Mycobacterium Tuberculosis CFP10 Monoclonal antibody specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mycobacterium Tuberculosis CFP10
Monoclonal
DyLight 350
10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant protein CFP10 M. tuberculosis H37RV.
0.2 mL
Primary
Store at 4°C in the dark.
IgG2b
ELISA, Sandwich ELISA
KFB42
50mM Sodium Borate
Mouse
Protein G purified
RUO
Bacteria