missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycobacterium tuberculosis Polyclonal Antibody, HRP

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173135
This item is not returnable.
View return policy
Description
The PA1-73135 antibody reacts with Mycobacterium tuberculosis. PA1-73135 has been successfully used in ELISA applications. The PA1-73135 antibody was raised against purified PPD from Mycobacterium tuberculosis.
Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
Specifications
| Mycobacterium tuberculosis | |
| Polyclonal | |
| HRP | |
| m tuberculosis; M. tuberculosis | |
| Purified PPD from Mycobacterium tuberculosis. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction