missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mycoplasma hominis Monoclonal Antibody (6502)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA533282

Product Code. 17115183

  • € 530.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

No cross reactivity with M. genitalium, M. pneumoniae and U. urealyticum.

Mycoplasma hominis is a species of bacteria that lacks a cell wall around their cell membranes. Mycoplasma hominis is one of the most frequently isolated mycoplasmas from the human genital tract and has been linked to pelvic inflammatory disease and preterm labor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mycoplasma hominis
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Bacteria
Antibody
IgG1
ELISA, Immunocytochemistry
6502
Unconjugated
mycoplasmen
Inactivated whole cell preparation of Mycoplasma hominis.
1 mL
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.