missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma pneumoniae MPN-083 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117544
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
MPN_083 is an uncharacterized lipoprotein.
Specifications
| Mycoplasma pneumoniae MPN-083 | |
| Polyclonal | |
| Unconjugated | |
| MPN_083; Uncharacterized lipoprotein MPN_083 | |
| Recombinant Mycoplasma pneumoniae Uncharacterized lipoprotein MPN_083 protein (22-242aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 3.79 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction