missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma pneumoniae Polyclonal Antibody, Biotin

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173159
This item is not returnable.
View return policy
Description
The PA1-73159 antibody reacts with Mycoplasma pneumoniae. PA1-73159 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73159 antibody was raised against purified M. pneumoniae.
Although they are the simplest and smallest self-replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The organism's affinity for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
Specifications
| Mycoplasma pneumoniae | |
| Polyclonal | |
| Biotin | |
| Bacterial pneumonia; M. pneumoniae; mycoplasmen | |
| Purified M. pneumoniae. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction