missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG5 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06128APC
This item is not returnable.
View return policy
Description
ABCG5 Polyclonal antibody specifically detects ABCG5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ABCG5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| ATP-binding cassette sub-family G member 5, ATP-binding cassette, sub-family G (WHITE), member 5, ATP-binding cassette, subfamily G, member 5, sterolin 1, sterolin-1, STSL | |
| Produced in rabbits immunized with E. coli-derived Human ABCG5 fragment. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 64240 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction