missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYD118 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06405-50ul
This item is not returnable.
View return policy
Description
MYD118 Polyclonal antibody specifically detects MYD118 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MYD118 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| DKFZp566B133, GADD45BETA, growth arrest and DNA damage-inducible protein GADD45 beta, growth arrest and DNA-damage-inducible, beta, MYD118DKFZP566B133, Myeloid differentiation primary response protein MyD118, Negative growth regulatory protein MyD118 | |
| Produced in rabbits immunized with E. coli-derived Human MYD118 fragment. | |
| 50 μg | |
| Cell Cycle and Replication | |
| 4616 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction