missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody (316628), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB29281Y
This item is not returnable.
View return policy
Description
MyD88 Monoclonal antibody specifically detects MyD88 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| MyD88 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 316628 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| E. coli-derived recombinant human MyD88, Met1-Pro296, Accession # Q99836 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction