missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody (4D6), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27369UV
This item is not returnable.
View return policy
Description
MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications
| MyD88 | |
| Monoclonal | |
| DyLight 350 | |
| Q99836 | |
| MYD88 | |
| A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 4D6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction