Learn More
MyD88 Antibody (OTI2B2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72842AF532
Product Code. 18183877
  • ONLINE EXCLUSIVE PRICE
    € 785.00
    € 742.35 / 0.10mL
    Save € 42.65
    5% Off
Description
MyD88 Monoclonal specifically detects MyD88 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MyD88
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
MYD88
Full length recombinant protein of human MyD88 (NP_002459) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG2a
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2B2
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Mouse
Affinity Purified
Cytokine Research, Immunology, Innate Immunity, Protein Kinase
4615.0
Store at 4°C in the dark.