missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYD88 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23230-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a cytosolic adapter protein that plays a central role in the innate and adaptive immune response. This protein functions as an essential signal transducer in the interleukin-1 and Toll-like receptor signaling pathways. These pathways regulate that activation of numerous proinflammatory genes. The encoded protein consists of an N-terminal death domain and a C-terminal Toll-interleukin1 receptor domain. Patients with defects in this gene have an increased susceptibility to pyogenic bacterial infections. Alternate splicing results in multiple transcript variants.Specifications
| MYD88 | |
| Polyclonal | |
| Unconjugated | |
| MYD88 | |
| MYD88, MYD88D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301059, 4615 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6Y1S1, Q99836 | |
| MYD88 | |
| MYD88 Fusion Protein Ag19770 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction