Learn More
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig
Specifications
Specifications
| CAS | 98474-59-0 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 1736.79 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 50 mg/mL (28.79 mM∣Need ultrasonic) |