Description
Myelin PLP Monoclonal antibody specifically detects Myelin PLP in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Myelin PLP |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL10622 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P60201, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |