Learn More
Myelin PLP Antibody (CL10622) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07989PCP
Product Code. 30300624
  • € 737.00 / 0.10mL
Description
Myelin PLP Monoclonal antibody specifically detects Myelin PLP in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Myelin PLP
Monoclonal
PerCP
HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2
This antibody was developed using a recombinant protein derived from P60201, with the exact immunogen sequence remaining proprietary.
0.1 mL
Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers
5354
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10622
PBS
Mouse
Protein A purified
RUO
Primary
Human, Rat