missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin Protein Zero Antibody (MPZ/7389), Novus Biologicals™
Mouse Monoclonal Antibody
€ 245.00 - € 462.00
Specifications
| Antigen | Myelin Protein Zero |
|---|---|
| Clone | MPZ/7389 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680115
|
Novus Biologicals
NBP3-23862-20ug |
20 μg |
€ 245.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678494
|
Novus Biologicals
NBP3-23862-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myelin Protein Zero Monoclonal antibody specifically detects Myelin Protein Zero in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Myelin Protein Zero | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Oligodendrocyte Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 4359 | |
| IgG2a κ | |
| Protein A or G purified |
| MPZ/7389 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0 | |
| Recombinant full-length human Myelin Protein Zero protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title