missing translation for 'onlineSavingsMsg'
Learn More

Myelin Protein Zero Antibody (MPZ/7390), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18576748 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18576748 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18576748 Supplier Novus Biologicals Supplier No. NBP324149AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myelin Protein Zero Monoclonal antibody specifically detects Myelin Protein Zero in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin Protein Zero
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MPZ/7390
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0
Host Species Mouse
Immunogen Recombinant full-length human Myelin Protein Zero protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4359
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.