missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid Cell Marker Antibody (SPM298), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34759JF646
This item is not returnable.
View return policy
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Myeloid Cell Marker | |
| Monoclonal | |
| Janelia Fluor 646 | |
| myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein | |
| Human peripheral blood mononuclear cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| SPM298 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction