Learn More
Invitrogen™ Myeloid Specific Antigen Monoclonal Antibody (OTI4H6)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526846
Product Code. 15986434
  • € 588.00 / 100µL
Description
Myeloid Specific Antigen Monoclonal Antibody for Western Blot, IHC (P)
This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Myeloid Specific Antigen
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P41218
MNDA
Human recombinant protein fragment corresponding to amino acids 37-321 of MNDA produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI4H6
Unconjugated
MNDA
Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135
Mouse
Affinity Chromatography
RUO
4332
-20°C, Avoid Freeze/Thaw Cycles