missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Myeloperoxidase/MPO Antibody (CL14991), Novus Biologicals™

Mouse Monoclonal Antibody

€ 317.00 - € 630.00

Specifications

Antigen Myeloperoxidase/MPO
Clone CL14991
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500
Applications Immunohistochemistry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043506
View Documents
Novus Biologicals™
NBP3-26846-100ul
100 μL
€ 630.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043590
View Documents
Novus Biologicals™
NBP3-26846-25ul
25 μL
€ 317.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications

Specifications

Myeloperoxidase/MPO
1 mg/mL
Immunohistochemistry, Western Blot
Unconjugated
Mouse
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
PBS, pH 7.2, 40% glycerol
4353
IgG3
Protein A purified
CL14991
Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500
Monoclonal
Purified
RUO
Human
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
This antibody was developed against a Synthetic Peptide of P05164. Actual immunogen sequence is proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.