missing translation for 'onlineSavingsMsg'
Läs mer

Myeloperoxidase/MPO Antibody (CL15002) [DyLight 680], Novus Biologicals Biologicals™

Produktkod. 30530800 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
0.1 mL
Förpackningsstorlek:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
30530800 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30530800 Leverantör Novus Biologicals Leverantörsnummer NBP344267FR

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Myeloperoxidase/MPO
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL15002
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P05164, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4353
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.