missing translation for 'onlineSavingsMsg'
Learn More

Myeloperoxidase/MPO Antibody (MPO/8290R), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 291.00 - € 519.00

Specifications

Antigen Myeloperoxidase/MPO
Clone MPO/8290R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18683084
View Documents
Novus Biologicals
NBP3-23861-100ug
100 μg
€ 519.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18638364
View Documents
Novus Biologicals
NBP3-23861-20ug
20 μg
€ 291.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Myeloperoxidase/MPO
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
10 mM PBS with 0.05% BSA
4353
IgG κ
Protein A or G purified
MPO/8290R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Recombinant fragment of human Myeloperoxidase/MPO protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.