missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (MPO/8997R), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24100APC
This item is not returnable.
View return policy
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Myeloperoxidase/MPO | |
| Monoclonal | |
| APC | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| Recombinant fragment of human Myeloperoxidase/MPO protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 4353 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MPO/8997R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction