missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (MPO421-8B2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41406B
This item is not returnable.
View return policy
Description
Myeloperoxidase/MPO Monoclonal specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Flow Cytometry, ELISA, Flow (Intracellular).
Specifications
| Myeloperoxidase/MPO | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MPO | |
| Human myeloperoxidase | |
| 0.1 mL | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 4353 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| MPO421-8B2 | |
| Flow Cytometry, ELISA, Flow (Intracellular) | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The mouse monoclonal antibody MPO421-8B2 recognizes human myeloperoxidase, a heme protein present in myeloblasts, neutrophils and monocytes. It is a marker of acute myelogenous leukemias and acute lymphoblastic leukemias. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction