missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase (MPO) Monoclonal Antibody (OTI3E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813904
This item is not returnable.
View return policy
Description
Myeloperoxidase (MPO) Monoclonal antibody specifically detects Myeloperoxidase (MPO) in Human samples. It is validated for ELISASpecifications
| Myeloperoxidase (MPO) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P05451 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI3E7 | |
| Unconjugated | |
| REG1A | |
| 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
| Human recombinant protein fragment corresponding to amino acids 23-166 of human PSP (NP_002900) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction