missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myeloperoxidase Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143017
This item is not returnable.
View return policy
Description
Additional Information: The immunizing peptide represents the N terminus of the mature protein. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
| Myeloperoxidase | |
| Polyclonal | |
| Unconjugated | |
| MPO | |
| 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4353 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P05164 | |
| MPO | |
| Peptide with sequence C-DKAYKERRESIKQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction