missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myeloperoxidase Recombinant Rabbit Monoclonal Antibody (PD00-20)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544664
This item is not returnable.
View return policy
Description
Myeloperoxidase Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
| Myeloperoxidase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P05164 | |
| MPO | |
| Synthetic peptide within Human Myeloperoxidase aa 600-700 (C terminal). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| PD00-20 | |
| Unconjugated | |
| MPO | |
| 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4353 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction