missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYF-5 Antibody (593128) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4027JF669
This item is not returnable.
View return policy
Description
MYF-5 Monoclonal antibody specifically detects MYF-5 in Human,Mouse samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| MYF-5 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, myogenic factor 5 | |
| E. coli-derived recombinant human MYF-5, Met1-Leu255, Accession # P13349 | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 593128 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4617 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction