missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYF6 Mouse anti-Human, Clone:5D7, Abnova™
Mouse monoclonal antibody raised against recombinant human MYF6.
Brand: Abnova MAB17730.100ug
This item is not returnable.
View return policy
Specifications
| MYF6 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry Immunocytochemistry Immunohistochemistry Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| MYF6 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 5D7 | |
| myogenic factor 6 (herculin) | |
| In PBS (0.05% sodium azide) | |
| MRF4, bHLHc4 | |
| Recombinant protein corresponding to amino acids 85-150 of human MYF6 from E. coli. | |
| 4618 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction