Learn More
Invitrogen™ MYF6 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA597990
Product Code. 16590812
  • € 461.00 / 100µg
Description
MYF6 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MYF6
Polyclonal
Unconjugated
Myf6
bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6
Rabbit
Protein G
RUO
17878, 25714, 4618
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
P15375, P19335, P23409
Myf6
Recombinant Human Myogenic factor 6 protein (26-232AA).
100 μg
Primary
Human, Mouse, Rat