Learn More
Invitrogen™ MYF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597990
This item is not returnable.
View return policy
Description
MYF6 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
Specifications
| MYF6 | |
| Polyclonal | |
| Unconjugated | |
| Myf6 | |
| bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 17878, 25714, 4618 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P15375, P19335, P23409 | |
| Myf6 | |
| Recombinant Human Myogenic factor 6 protein (26-232AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |