Learn More
MYF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11754-1-AP-150UL
 View more versions of this product
Product Code. 16849043
  • € 410.00 / 150µL
Description
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
Specifications
Specifications
MYF6
Polyclonal
Unconjugated
MYF6
bHLHc4, MRF4, Myf 6, MYF6, Myogenic factor 6, myogenic factor 6 (herculin)
Rabbit
Antigen Affinity Chromatography
RUO
17878, 25714, 4618
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15375, P19335, P23409
Myf6
MYF6 Fusion Protein Ag2350
150 μL
Primary
Human, Mouse, Rat