missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH10 Polyclonal Antibody

MYH10 Polyclonal Antibody
Brand: Invitrogen PA578065
This item is not returnable.
View return policy
Description
MYH10 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| MYH10 | |
| Polyclonal | |
| Unconjugated | |
| MYH10 | |
| 5730504C04Rik; 9330167F11Rik; cellular myosin heavy chain, type B; flectin; Fltn; MCH-B; MCH-B(B2); mKIAA3005; MYH10; Myhn2; Myhn-2; Myosin heavy chain 10; myosin heavy chain 10, non-muscle; myosin heavy chain, non-muscle IIb; myosin heavy chain, nonmuscle type B; myosin heavy chain, nonmuscular, type B; myosin IIB; myosin, heavy chain 10, non-muscle; myosin, heavy polypeptide 10, non-muscle; myosin-10; NMHC II-B; NMHC-B; NMHCII-B; NMMHC IIB; NMMHC II-b; NMMHCB; NMMHC-B; NMMHC-IIB; nonmuscle myosin; Non-muscle myosin heavy chain B; nonmuscle myosin heavy chain IIB; non-muscle myosin heavy chain IIb; nonmuscle myosin heavy chain II-B; nonmuscle myosin heavy chain-B; nonmuscle myosin II heavy chain-B; OTTHUMP00000165799; SMemb | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.72 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P35580 | |
| MYH10 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Human MYH10. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction