Learn More
MYH14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20716-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to MYH14.
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.Specifications
| MYH14 | |
| Polyclonal | |
| Unconjugated | |
| MYH14 | |
| MHC16, MYH14, myosin, Myosin 14, Myosin heavy chain 14, myosin, heavy chain 14, NMHC II C, non-muscle II c | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 308572, 71960, 79784 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6URW6, Q7Z406 | |
| Myh14 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |