missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYH15 Polyclonal Antibody

Product Code. 17894600
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17894600 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17894600 Supplier Invitrogen™ Supplier No. PA5140088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGEAAAFLRR SEAELLLLQA TALDGKKKCW IPDGENAYIE AEVKGSEDDG TVIVETADGE SLSIKEDKIQ QMNPPEFE Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 69%.

Muscle contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH15
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYH15
Gene Accession No. Q9Y2K3
Gene Alias KIAA1000; MYH15; Myosin heavy chain 15; myosin, heavy chain 15; myosin, heavy polypeptide 15; myosin-15
Gene Symbols MYH15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYH15. Recombinant protein control fragment (Product #RP-108948).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22989
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.