Learn More
MYH6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Product Code. 18019375 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18019375 20 μL 20µL
18020726 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18019375 Supplier Proteintech Supplier No. CL4882228120UL
Rabbit Polyclonal Antibody
The MYH6 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH6. This antibody recognizes human, mouse, rat antigen. The MYH6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Antigen MYH6
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols alpha MHC, ASD3, MYH6, MYHC, MyHC alpha, MYHCA, Myosin 6, Myosin heavy chain 6
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 4624
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG