missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MYH7 Antibody (MYH7/9183), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26826-20ug

 View more versions of this product

Product Code. 30043502

  • € 232.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MYH7 Monoclonal antibody specifically detects MYH7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MYH7
Monoclonal
0.2 mg/mL
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
cardiac muscle beta isoform, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM
Recombinant fragment (around aa1150-1350) of human MYH7 protein (exact sequence is proprietary)
20 μg
Cell Biology, Cytoskeleton Markers, Signal Transduction
4625
Store at 4°C.
IgG2b κ
Immunohistochemistry (Paraffin)
MYH7/9183
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.