missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MYH7 Antibody (MYH7/9184), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26827-100ug
This item is not returnable.
View return policy
Description
MYH7 Monoclonal antibody specifically detects MYH7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MYH7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| cardiac muscle beta isoform, CMD1SMGC138376, CMH1, DKFZp451F047, MGC138378, MPD1, MYHCB, MyHC-beta, myhc-slow, myopathy, distal 1, myosin heavy chain (AA 1-96), myosin, heavy chain 7, cardiac muscle, beta, myosin, heavy polypeptide 7, cardiac muscle, beta, myosin-7, rhabdomyosarcoma antigen MU-RMS-40.7A, SPMD, SPMM | |
| Recombinant fragment (around aa1150-1350) of human MYH7 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4625 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| MYH7/9184 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction