missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYH7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143722
This item is not returnable.
View return policy
Description
The antibody detects a 215 kDa* protein corresponding to the apparent molecular mass of MYH7B on SDS-PAGE immunoblots of mouse extraocular muscle and brain.
Myosin cardiac muscle beta chain or MYH7B is believed to be involved in cardiac slow-twitch myosin contraction.
Specifications
| MYH7B | |
| Polyclonal | |
| Unconjugated | |
| MYH7B | |
| MYH7B | |
| A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the hinge region from mouse MYH7B/MHC14. This sequence is highly conserved in rat and human MYH7B, and has less than 50% identity with other MYH family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| A2AQP0, A7E2Y1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 311570, 57644, 668940 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction