missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH8-Specific Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MYH8-Specific |
|---|---|
| Concentration | 200 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237164
|
Proteintech
18988-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234944
|
Proteintech
18988-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MYH8-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH8-Specific. This antibody recognizes mouse antigen. The MYH8-Specific antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| MYH8-Specific | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| MyHC-peri; MyHC-pn | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 200 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 4626 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title