missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH9, Mouse anti-Human, Clone: 3C7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004627-M01.50ug
This item is not returnable.
View return policy
Description
Sequence: RLKQLKRQLEEAEEEAQRANASRRKLQRELEDATETADAMNREVSSLKNKLRRGDLPFVVPRRMARKGAGDGSDEEVDGKADGAEAKPAESpecifications
| MYH9 | |
| Monoclonal | |
| Unconjugated | |
| myosin, heavy chain 9, non-muscle | |
| DFNA17/EPSTS/FTNS/MGC104539/MHA/NMHC-II-A/NMMHCA | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3C7 | |
| In 1x PBS, pH 7.4 | |
| NM_002473.5 | |
| MYH9 | |
| MYH9 (NP_002464.1, 1871 a.a. ∼ 1960 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4627 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction